Anti-TLR7 Mouse Monoclonal Antibody [clone: 4G6]

供应商: Novus Biologicals
NOVUNBP2-27332EA 0 CNY
NOVUNBP2-27332
Anti-TLR7 Mouse Monoclonal Antibody [clone: 4G6]
抗体
Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: monoclonal
Clone: 4G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR