Anti-PTEN Mouse Monoclonal Antibody [clone: 3E7]

供应商: Abnova
ABNOH00005728-M02EA 0 CNY
ABNOH00005728-M02
Anti-PTEN Mouse Monoclonal Antibody [clone: 3E7]
抗体
Type: Primary
Antigen: PTEN
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR