Anti-CD70 Mouse Polyclonal Antibody

供应商: Abnova
ABNOH00000970-B01PEA 0 CNY
ABNOH00000970-B01P
Anti-CD70 Mouse Polyclonal Antibody
抗体
Type: Primary
Antigen: CD70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR