Beakers, Berzelius, Borosilicate glass, KIMBLE®
供应商: DWK Life Sciences
KIML14030-200EA 0 CNY
KIML14030-400 KIML14030-200 KIML14020-100
Beakers, Berzelius, Borosilicate glass, KIMBLE®
烧杯
These Berzelius beakers, KIMAX® borosilicate glass, are ideally used for performing titrations and have slightly flared top and a thick reinforcing bead.
  • With or without spout
  • Very good chemical resistance
  • High thermal shock resistance
  • Excellent mechanical strength and durability
All sizes have a durable matte finish area for marking with an ordinary pencil and also it has white scales for reading the volumes easily.