The Stealth goggles combine modern design, technology, materials and high-performance lens coatings to create the highest levels of worker comfort and acceptance.
- Lens can be replaced easily and quickly
- Indirect ventilation reduces fogging and improves vision
- Quick adjust headband
- Rx insert available
Protect against dust, mist, chemical splashes and impacts. Durable polypropylene frame combined with soft, flexible elastomer conforms to facial contours for a comfortable seal.
Weight: 75,2 g
Certifications: ANSI Z87+ and CSA Z94.3 high impact protector standards.