Chemiluminiscent imager, ImageQuant™ LAS 500
供应商: Cytiva
GEHE29-0050-63EA 0 CNY
GEHE29-0050-63
Chemiluminiscent imager, ImageQuant™ LAS 500
成像系统
ImageQuant™ LAS 500 is a cooled CCD imager for imaging of chemiluminescent Western blots, fluorescent protein and DNA gel stains, and white light imaging of colorimetric stains and markers.
  • Sensitivity: detect picogram levels of sample with chemiluminiscence
  • Automatic overlay function: simultaneously image chemiluminescent samples and coloured molecular weight markers
  • Ready-to-operate in less than 5 min with the rapid cooling camera
  • Compact integrated design works independently of a dedicated PC with flexible image storage
  • Flexible storage of image on a USB memory stick, on the system or in a network connection folder
ImageQuant™ LAS 500 is a practical and affordable solution for individual researchers or laboratories that are either looking for their own imager or need extra imaging capacity. The 300×280 mm footprint has the same area as a typical laptop computer, which allows the imager to fit neatly on almost any lab bench without the need to connect ancillary equipment.
The CCD camera is ready for use in minutes, and advanced Peltier cooling and binning significantly reduce noise levels, increasing both the sensitivity and linearity of the system. It is designed for chemiluminescence detection with regard to sensitivity and dynamic range.
In addition, the system has three LED light sources; a combined blue epi-/UV, epi-light and a white epi-light. The combined blue epi-/UV and epi-light allows imaging of fluorescence signals above 560 nm. This feature makes the camera well suited for DNA stain imaging with SYBR™ Green and ethidium bromide (EtBr) as well as with common total protein stain dyes. The white epi-light source (470 to 635 nm) is for documentation of colorimetric markers and stains, such as Coomassie™ Blue or silver.
4,8 orders of magnitude.
Delivery information: Supplied with touchscreen, sample chamber, USB port, camera and light sources.