Industrial bench scales, FOB series
供应商: Kern & Sohn
KERNFOB6K2EA 0 CNY
KERNFOB3K1 KERNFOB6K2 KERNFOB1.5K0.5
Industrial bench scales, FOB series
天平
This series of portable scales with stainless steel housing is designed for use in food and packaging applications. The scales are smooth, easy to clean and give increased protection against humidity via the waterproof silicone sealing of the load cell, electronics and soldering joints.
Certifications: Please note: If a calibration certificate is required, this should be requested when ordering the balance, as it can not be issued retrospectively.