Precision balances, PLS/PLJ series
供应商: Kern & Sohn
KERNPLJ2000-3AEA 0 CNY
KERNPLJ2000-3A KERNPLJ420-3F KERNPLS420-3F KERNPLS4200-2F KERNPLJ4200-2F KERNPLS6200-2A KERNPLS1200-3A KERNPLS720-3A
Precision balances, PLS/PLJ series
天平
Precision balances designed with ergonomically optimised keypad for left and right handed users.
  • PLJ: Automatic internal adjustment
  • PLS: Adjusting program CAL for quick setting of the balance accuracy
Draft shield standard on all models with readability= 0,001 g. Removable metal cover with pipette opening.
Balances with F and GM suffix: Weighing principle is strain gauge
Balances with A and CM suffix: Weighing principle is force compensation
Certifications: Please note: If a calibration certificate is required, this should be requested when ordering the balance, as it can not be issued retrospectively.