(R)-(+)-2-氨基-3-苯基-1-丙醇 98%

供应商: Thermo Scientific
Danger

Synonyms: D-苯丙氨醇

ACRO417340100EA 0 CNY
ACRO417340100 ACRO417340010
(R)-(+)-2-氨基-3-苯基-1-丙醇 98%
(R)-(+)-2-氨基-3-苯基-1-丙醇
分子式: C₉H₁₃NO
分子量: 151,2 g/mol
溶点: 90…94 °C
存储条件: Ambient
MDL编号: MFCD00064298
CAS号: 5267-64-1
EINECS: 226-086-1
UN号: 3259
ADR: 8,III

现在订购

产品技术规格

Appearance White to yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 90 to 94 °C
HPLC ≥97.5 %
Specific optical rotation +21° to +25° (22 °C, 589 nm) (c=1.2, 1 N HCl)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR