二硫化碳 ≥99.9% (通过GC,校正水含量), AR® ACS, Macron Fine Chemicals™
供应商: Avantor Performance Materials
|
Danger
|
MACR8831-03EA
0
CNY
MACR8831-03
二硫化碳 ≥99.9% (通过GC,校正水含量), AR® ACS, Macron Fine Chemicals™
二硫化碳
Suitable for NIOSH Procedure P and Cam 127.
分子式:
CS₂ 分子量: 76,14 g/mol 沸点: 46,2 °C (1013 hPa) 溶点: –111,6 °C 密度: 1,2632 g/cm³ (20 °C) 闪点: –30 °C 存储条件: Ambient |
MDL编号:
MFCD00011321 CAS号: 75-15-0 EINECS: 200-843-6 UN号: 1131 ADR: 3,I Merck Index: 13,01821 |
产品技术规格
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CS₂) (by GC, corrected for water) | ≥ 99.9 % |
ACS - Color (APHA) | ≤ 10 |
ACS - Residue after Evaporation | ≤ 0.002 % |
ACS - Hydrogen Sulfide (H₂S) | Passes Test |
ACS - Sulfur Dioxide (SO₂) | Passes Test |
ACS - Water (by coulometry) | ≤ 0.05 % |
ACS - Benzene (C₆H₆) | ≤ 1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...