The senION combination ammonia gas-sensing ion selective electrode (ISE) features a replaceable membrane module, refillable electrolyte and fixed cable with BNC connector. The electrode comes with three ammonia membrane modules and a bottle of electrode filling solution.
- Refillable outer body, an easy to maintain modular membrane design, and a large annular reference junction for maximum surface area between sample and reference electrolyte provide a fast and stable measurement
- Innovative menu designs and simple keypads for intuitive operation
- Rugged and durable: Probes are designed to withstand the rigors of outdoor testing and frequent use
- Flexible: Can be used in the lab or in the field
Ammonia ISE is compatible with sensION, H-series, and sensION+ systems. ISE compatible sensION+ meters are the MM340 GLP pH/ISE laboratory meter or the sensION+ MM374 laboratory multimeter.
Delivery information: Electrode is supplied with three ammonia membrane modules, electrode internal fill solution (50 ml), and user manual.