1,4-二氧六环

供应商: LOCAL DEALS CHINA
Danger

CHLCR00273320116EA 0 CNY
CHLCR00273320116
1,4-二氧六环
1,4-二氧六环
分子式: C₄H₈O₂
分子量: 88,11 g/mol
沸点: 101,1 °C (1013 hPa)
溶点: 11,8 °C
密度: 1,03 g/cm³ (20 °C)
闪点: 12 °C
存储条件: Ambient
MDL编号: MFCD00006571
CAS号: 123-91-1
EINECS: 204-661-8
UN号: 1165
ADR: 3,II
Merck Index: 13,03330
https://cn.vwr.com/stibo/web/std.lang.all/38/27/773827.jpg

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR