Soaps, general purposes, NovaDyne™
供应商: Micronova
NovaDyne™
MICRIC-110EA 0 CNY
MICRIC-110
Soaps, general purposes, NovaDyne™
护肤用品 肥皂和皮肤清洁剂
General purpose lathering liquid soap for use in commercial settings.
  • Contains emollients
  • pH balanced so skin is left feeling smooth and deodorized
  • Synthetic formulation leaving hands free of any residue
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904