1,1,1,3,3,3-六氟异丙基异丁烯酸酯 99% stabilised
供应商: Thermo Scientific
|
Danger
|
ACRO308930050EA
0
CNY
ACRO308930050
1,1,1,3,3,3-六氟异丙基异丁烯酸酯 99% stabilised
1,1,1,3,3,3-六氟异丙基异丁烯酸酯
分子式:
C₇H₆F₆O₂ 分子量: 236,12 g/mol 闪点: 14 °C |
MDL编号:
MFCD00040105 CAS号: 3063-94-3 EINECS: 221-309-9 UN号: 3272 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3300 to 1.3320 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | Exact type and amount of stabilizer on the CoA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...