1,2-二甲氧基乙烷
供应商: LOCAL DEALS CHINA
|
Danger
|
Synonyms:
单甘醇二甲醚
, 1,2-二甲氧基乙烷
, mono-Glyme
, 乙二醇二甲醚
CHLCR00928930100EA
0
CNY
CHLCR00928930100
CHLCR00928930124
1,2-二甲氧基乙烷
1,2-二甲氧基乙烷
分子式:
C₄H₁₀O₂ 分子量: 90,12 g/mol 沸点: 85 °C (1013 hPa) 溶点: –69 °C 密度: 1,113 g/cm³ (20 °C) 闪点: –6 °C 存储条件: Ambient |
MDL编号:
MFCD00008502 CAS号: 110-71-4 EINECS: 203-794-9 UN号: 2252 ADR: 3,II Merck Index: 13,03251 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...