酚酞

供应商: LOCAL DEALS CHINA
Danger

CHLCR00220990133EA 0 CNY
CHLCR00220990133
酚酞
酚酞
分子式: C₂₀H₁₄O₄
分子量: 318,33 g/mol
溶点: 258…262 °C
密度: 1,3 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00005913
CAS号: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 13,07325
https://cn.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR