HPLC columns, NUCLEOSIL® C18 HD
供应商: Macherey-Nagel
NUCLEOSIL®
MANA720296.46EA 0 CNY
MANA721072.30 MANA720280.40 MANA720191.40 MANA720193.46 MANA720296.40 MANA720280.46 MANA720191.46 MANA720294.46 MANA721196.30 MANA720296.46
HPLC columns, NUCLEOSIL® C18 HD
色谱柱
NUCLEOSIL® C18 HD is an octadecyl modified silica phase with a high density coverage, using for HPLC.
  • Robust spherical silica gel
  • Strongly hydrophobic, endcapped phase for RP chromatography
  • Separation mechanism based on hydrophobic interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).