Abaque™ Peristaltic Natural Rubber Replacement Hose
供应商: MASTERFLEX PROPRIETARY
75810-21
MFLA75810-32EA 0 CNY
MFLA75810-32 MFLA75810-43 MFLA75810-48 MFLA75810-83 MFLS75810-21 MFLA75810-64
Abaque™ Peristaltic Natural Rubber Replacement Hose
胶管 泵管
Highly resilient with excellent abrasion resistance and strength.
  • All listed hoses in this ad are made from Natural Rubber.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Abaque engineers / manufactures their hoses from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy,More Product Information