Abaque HDN40 Replacement Hose, Natural Rubber, 1.5" ID x 2.48" OD; 49.21" L
供应商: MASTERFLEX PROPRIETARY
VWR Item#MFLA75810-48
Your Organization's Price:
正在获取
Highly resilient with excellent abrasion resistance and strength.
  • All listed hoses in this ad are made from Natural Rubber.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Abaque engineers / manufactures their hoses from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy, longevity, and optimal compression. <br /><br />Ideal for applications where the following properties are needed:<br />Highly resilient, excellent abrasion resistance, the most common hose in peristaltic markets. Generally resistant with lightly corrosive chemical applications, abrasive slurries, etc.