Extensive refractive index standard range from (1,34325-1,65808 ŋD)
A complete range of 25 standards solvent-based and sucrose-stabilised at 20°C
These 25 refractive index standards can be used with any brand of refractometer.
  • One of the most extensive range in the marketplace
  • Presented in a convenient high quality 15 ml dropper bottle
  • Uncertainty of measurement up to ±0,00014 ŋD units
  • Consistency of product: Independent, traceable, certified
  • Certificates of Analysis and Safety Data Sheets available online