Secuflow by Waldner
Innovative technology:
With Waldner laboratory fume cupboards, whatever their design, you are guaranteed maximum safety, operational comfort and ergonomics with outstanding economic efficiency.
Many years of design know-how and scientific research has gone into the development of these fume cupboards.
Waldner Fume Cupboards
available as:
- Bench mounted fume cupboards
- Low ceiling bench mounted fume cupboards
- Low level fume cupboards & walk-in fume cupboards
- Secuflow
- Fume cupboards with side panels
- Special application fume cupboards
- Filter fume cupboards
- Radio-isotope fume cupboards
- Mobile fume cupboards
All Waldner fume cupboards fulfil the requirements of all customary
standards and regulations and are certified according the new EN 14175.
- Safety via the patented intake airflow profile on the front edge of the fume cupboard worktop
- Clear view of all processes inside the fume cupboard due to glazed panel in top of cupboard
- Maximum utilisation of working width thanks to patented side posts only 25 mm in width
- Automatic closure of the sash, photo-electric barrier and motion detector are incorporated in the overall system to optimum effect
- Secuflow extraction with integrated supportive flow technology offers maximum economic efficiency and safety with much lower exhaust air