Experience Tangible Perfection at Your Lab With Acuumax Centrifuge Tubes
PFACT Centrifuge Tubes are made by USFDA approved virgin PP raw material ensuring compatibility for critical applications.
- Wide writing patch with calibrated graduation
- Swiss made diamond mold finishing, specially designed trace metal free tube caps.
- Withstand G-Force with upto 20000
- Super leak proof seal design