VWR® Petri Dishes with Lid, Soda-lime Glass
供应商: VWR Collection
VWRI391-0652EA 630 CNY
VWRI391-0652 VWRI391-0660 VWRI391-0665 VWRI391-0668 VWRI391-0671 VWRI391-0673 VWRI391-0674 VWRI391-0707 VWRI391-0708
VWR® Petri Dishes with Lid, Soda-lime Glass
培养皿 培养皿
Glass Petri dishes are ideal for cultivating bacteria, fungi, and other microorganisms, as well as for cell cultures in research and diagnostics.
  • Durable, reusable, and easy to clean
  • A sustainable choice
  • Chemically inert glass eliminates unwanted reactions with cultures or samples
  • Transparent glass makes it easier to observe growth and development
Glass Petri dishes are durable, reusable, and they provide a sterile environment for various scientific applications.