3-Ethoxy-4-methoxybenzaldehyde ≥98.0% (by GC)
供应商: TCI
Synonyms: Apremilast Impurity 28
E0835-25G E0835-5G
TCIAE0835-25GEA 0 CNY
TCIAE0835-25G TCIAE0835-5G
3-Ethoxy-4-methoxybenzaldehyde ≥98.0% (by GC)
3-Ethoxy-4-methoxybenzaldehyde
分子式: C₁₀H₁₂O₃
沸点: 155°C
MDL编号: MFCD00010128
CAS号: 1131-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8df1b8841cd5d0847ea1a270f1435d0a12976dda&vid=8df1b8841cd5d0847ea1a270f1435d0a12976dda&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C