3,4′-二氨基二苯醚 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-(4-Aminophenoxy)aniline, 3,4'-Oxydianiline

D1924-25G D1924-5G
TCIAD1924-25GEA 0 CNY
TCIAD1924-25G TCIAD1924-5G
3,4′-二氨基二苯醚 ≥97.0% (by GC, titration analysis)
3,4′-二氨基二苯醚
分子式: C₁₂H₁₂N₂O
分子量: 200.2365 g/mol
MDL编号: MFCD00036097
CAS号: 2657-87-6
EINECS: 220-190-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=545d746ebd9fd6094eb8e9ccc0d0002f3a4e8b56&vid=545d746ebd9fd6094eb8e9ccc0d0002f3a4e8b56&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR