二甲氧基乙酸甲酯 ≥95.0% (by GC)

供应商: TCI

Synonyms: 2,2-二甲氧基乙酸甲酯

D0503-10ML D0503-25ML
TCIAD0503-10MLEA 0 CNY
TCIAD0503-10ML TCIAD0503-25ML
二甲氧基乙酸甲酯 ≥95.0% (by GC)
二甲氧基乙酸甲酯
分子式: C₅H₁₀O₄
沸点: 67°C
闪点: 67°C
MDL编号: MFCD00008484
CAS号: 89-91-8
EINECS: 201-950-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4005b8cff02438214bc0819c6754d8fd7a4b9538&vid=4005b8cff02438214bc0819c6754d8fd7a4b9538&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR