异丁酸异戊酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Isoamyl isobutyrate
I0676-25G
I0676-500G
TCIAI0676-25GEA
0
CNY
TCIAI0676-25G
TCIAI0676-500G
异丁酸异戊酯 ≥98.0% (by GC)
异丁酸异戊酯
分子式:
C₉H₁₈O₂ 沸点: 169°C 闪点: 52°C |
MDL编号:
MFCD00053719 CAS号: 2050-01-3 EINECS: 218-078-1 UN号: 2620 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...