2,6-二乙酰基吡啶 ≥98.0% (by GC)

供应商: TCI
Warning

D2377-25G D2377-5G
TCIAD2377-25GEA 0 CNY
TCIAD2377-25G TCIAD2377-5G
2,6-二乙酰基吡啶 ≥98.0% (by GC)
2,6-二乙酰基吡啶
分子式: C₉H₉NO₂
沸点: 126°C
MDL编号: MFCD00006304
CAS号: 1129-30-2
EINECS: 214-442-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96a1b4ee04c5f6b5e3387ec2a073c194134faa15&vid=96a1b4ee04c5f6b5e3387ec2a073c194134faa15&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR