Thionicotinamide 99%

供应商: Thermo Fisher Scientific

138800250 138801000
ACRO138800250EA 0 CNY
ACRO138800250 ACRO138801000
Thionicotinamide 99%
Thionicotinamide
分子式: C₆H₆N₂S
溶点: 185…190 °C
闪点: 160 °C
存储条件: Ambient
MDL编号: MFCD00006399
CAS号: 4621-66-3
EINECS: 225-036-6

现在订购

产品技术规格

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Melting point 183 to 190 °C (Dec)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR