3,5-双(三氟甲基)苄基氯 ≥97.0% (通过GC)
供应商: TCI
|
Danger
|
B3641-25G
B3641-5G
TCIAB3641-25GEA
0
CNY
TCIAB3641-25G
TCIAB3641-5G
3,5-双(三氟甲基)苄基氯 ≥97.0% (通过GC)
3,5-双(三氟甲基)苄基氯
分子式:
C₉H₅ClF₆ 沸点: 84°C 闪点: 71°C |
MDL编号:
MFCD00009906 CAS号: 75462-59-8 EINECS: 278-216-1 UN号: 3265 ADR: 8,II |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...