N-叔丁基-3,5-二甲基苯胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
N-(1,1-二甲基乙基)-3,5-二甲基苯胺
B2903-1G
B2903-5G
TCIAB2903-1GEA
0
CNY
TCIAB2903-1G
TCIAB2903-5G
N-叔丁基-3,5-二甲基苯胺 ≥98.0% (by GC)
N-叔丁基-3,5-二甲基苯胺
分子式:
C₁₂H₁₉N 沸点: 70°C 存储条件: Refrigerator |
MDL编号:
MFCD07440135 CAS号: 110993-40-3 |
产品技术规格
Appearance | Colorless to Brown to Dark purple clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...