2,5-Dichlorosulphanilic acid ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2,5-Dichloroaniline-4-sulphonic acid
D0415-25G
D0415-500G
TCIAD0415-25GEA
0
CNY
TCIAD0415-25G
TCIAD0415-500G
2,5-Dichlorosulphanilic acid ≥98.0% (by HPLC, titration analysis)
2,5-Dichlorosulphanilic acid
分子式:
C₆H₅Cl₂NO₃S |
MDL编号:
MFCD00025159 CAS号: 88-50-6 EINECS: 201-836-0 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot DMF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...