异丁酸2-苯氧乙酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: 2-甲基丙酸苯氧乙酯

P1822-25G P1822-500G
TCIAP1822-25GEA 0 CNY
TCIAP1822-25G TCIAP1822-500G
异丁酸2-苯氧乙酯 ≥98.0% (by GC)
异丁酸2-苯氧乙酯
分子式: C₁₂H₁₆O₃
沸点: 127°C
闪点: 113°C
MDL编号: MFCD00027363
CAS号: 103-60-6
EINECS: 203-127-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=478fb0ee33ce3ee28b21e84f5eaadb1fa88d1693&vid=478fb0ee33ce3ee28b21e84f5eaadb1fa88d1693&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR