2-乙酰氧基异丁酰氯 ≥97.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-乙酸基-2-甲基丙酰氯, 2-乙酰氧基异丁酰氯
A1120-25G
A1120-5G
TCIAA1120-25GEA
0
CNY
TCIAA1120-25G
TCIAA1120-5G
2-乙酰氧基异丁酰氯 ≥97.0% (by titrimetric analysis)
2-乙酰氧基异丁酰氯
分子式:
C₆H₉ClO₃ 分子量: 164.59 g/mol 沸点: 70°C 闪点: 68°C |
MDL编号:
MFCD00000708 CAS号: 40635-66-3 EINECS: 255-016-2 UN号: 3265 ADR: 8 II |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...