N,N′-二仲丁基对苯二胺 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: N,N'-Di-sec-butyl-1,4-phenylenediamine, N1,N4-Di-sec-butylbenzene-1,4-diamine

D2268-25ML D2268-500ML
TCIAD2268-25MLEA 0 CNY
TCIAD2268-25ML TCIAD2268-500ML
N,N′-二仲丁基对苯二胺 ≥98.0% (by GC)
N,N′-二仲丁基对苯二胺
分子式: C₁₄H₂₄N₂
沸点: 159°C
闪点: 180°C
MDL编号: MFCD00043658
CAS号: 101-96-2
EINECS: 202-992-2
UN号: 2922
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=838daf2dcd89569e550979c65bc31c30a8653a7d&vid=838daf2dcd89569e550979c65bc31c30a8653a7d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR