D-苹果酸二甲酯 ≥98.0% (by GC)
供应商: TCI
Synonyms:
(R)-2-羟基琥珀酸二甲酯, Dimethyl D-malate
M1347-25G
M1347-5G
TCIAM1347-25GEA
0
CNY
TCIAM1347-25G
TCIAM1347-5G
D-苹果酸二甲酯 ≥98.0% (by GC)
D-苹果酸二甲酯
分子式:
C₆H₁₀O₅ |
MDL编号:
MFCD00066216 CAS号: 70681-41-3 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific rotation [a]20/D | +5.0 to +8.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...