Purpald ≥95.0% (by titrimetric analysis) for determination of aldehydes

供应商: TCI
Warning

Synonyms: 4-氨基-5-肼基-1,2,4-三唑-3-硫醇, 4-氨基-3-肼基-5-巯基-1,2,4-三唑

A0956-25G A0956-5G
TCIAA0956-25GEA 0 CNY
TCIAA0956-25G TCIAA0956-5G
Purpald ≥95.0% (by titrimetric analysis) for determination of aldehydes
Purpald
分子式: C₂H₆N₆S
溶点: 240°C
MDL编号: MFCD00003098
CAS号: 1750-12-5
EINECS: 217-135-8
UN号: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b6897412c8b9ba5940e87d99168c92c3e15453a&vid=b04fd48db1cf89911f99100ff5dd296032081fc8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR