2,5-二硫基-1,3,4-噻二唑 ≥98.0% (通过滴定分析)

供应商: TCI
Danger

D0622-25G D0622-500G
TCIAD0622-25GEA 0 CNY
TCIAD0622-25G TCIAD0622-500G
2,5-二硫基-1,3,4-噻二唑 ≥98.0% (通过滴定分析)
分子式: C₂H₂N₂S₃
MDL编号: MFCD00003103
CAS号: 1072-71-5
EINECS: 214-014-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=250782d89e8867bbdcea471131a5bdff5f1f3891&vid=250782d89e8867bbdcea471131a5bdff5f1f3891&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Sodium hydroxide solution almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR