4-(4-氯苯基)-4-羟基哌啶 ≥97.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
4-(4-氯苯基)-4-羟基哌啶, 4-(4-Chlorophenyl)-4-hydroxypiperidine, 4-(4-氯苯基)-4-哌啶醇
C1291-25G
C1291-5G
TCIAC1291-25GEA
0
CNY
TCIAC1291-25G
TCIAC1291-5G
4-(4-氯苯基)-4-羟基哌啶 ≥97.0% (by HPLC, titration analysis)
4-(4-氯苯基)-4-羟基哌啶
分子式:
C₁₁H₁₄ClNO 分子量: 211.69 g/mol |
MDL编号:
MFCD00006001 CAS号: 39512-49-7 EINECS: 254-479-8 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 138.0 to 142.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...