2-硝基噻吩 ≥80.0% (通过GC) (contains 3-Nitrothiophene)
供应商: TCI
|
Warning
|
N0407-25G
N0407-5G
TCIAN0407-25GEA
0
CNY
TCIAN0407-25G
TCIAN0407-5G
2-硝基噻吩 ≥80.0% (通过GC) (contains 3-Nitrothiophene)
2-硝基噻吩
分子式:
C₄H₃NO₂S 沸点: 224°C 密度: 1,3644 g/cm³ (42.6 °C) 存储条件: Ambient |
MDL编号:
MFCD00005425 CAS号: 609-40-5 EINECS: 210-190-9 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 80.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...