甲基丙烯酸月桂酯 ≥97.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: 甲基丙烯酸十二烷基酯

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M0083-25ML M0083-500ML
TCIAM0083-25MLEA 0 CNY
TCIAM0083-25ML TCIAM0083-500ML
甲基丙烯酸月桂酯 ≥97.0% (by GC) stabilised
甲基丙烯酸月桂酯
分子式: C₁₆H₃₀O₂
沸点: 167°C
闪点: 143°C
MDL编号: MFCD00008972
CAS号: 142-90-5
EINECS: 205-570-6
UN号: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24f2dab29e9b6568e0f709879ede4b8c5ae9ffff&vid=24f2dab29e9b6568e0f709879ede4b8c5ae9ffff&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR