1,5-二氮杂双环[4.3.0]壬-5-烯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: DBN

D1313-10ML D1313-25ML D1313-250ML
TCIAD1313-10MLEA 0 CNY
TCIAD1313-10ML TCIAD1313-25ML TCIAD1313-250ML
1,5-二氮杂双环[4.3.0]壬-5-烯 ≥98.0% (by GC)
1,5-二氮杂双环[4.3.0]壬-5-烯
分子式: C₇H₁₂N₂
分子量: 124.19 g/mol
沸点: 108°C
闪点: 98°C
存储条件: Ambient
MDL编号: MFCD00005554
CAS号: 3001-72-7
EINECS: 221-087-3
UN号: 3267
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79e44ff326e1d9c3f54bd11dcfff5fd87502d242&vid=79e44ff326e1d9c3f54bd11dcfff5fd87502d242&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR