4,4'-偶氮二苯甲醚 ≥97.0% (by GC)

供应商: TCI

Synonyms: 4,4'-二甲氧基氧化偶氮苯, 4,4′-氧化偶氮二苯甲醚

A0554-25G A0554-5G
TCIAA0554-25GEA 0 CNY
TCIAA0554-25G TCIAA0554-5G
4,4'-偶氮二苯甲醚 ≥97.0% (by GC)
4,4'-偶氮二苯甲醚
分子式: C₁₄H₁₄N₂O₃
MDL编号: MFCD00008400
CAS号: 1562-94-3
EINECS: 216-348-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=079c9059d4f22004a3f91d3e2c6413341a0802cf&vid=079c9059d4f22004a3f91d3e2c6413341a0802cf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR