苯并[b]噻吩-2-基硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 1-苯并噻吩-2-硼酸, 苯并噻吩-2-基硼酸, 苯并噻吩-2-硼酸

B2893-1G B2893-25G B2893-5G
TCIAB2893-1GEA 0 CNY
TCIAB2893-1G TCIAB2893-25G TCIAB2893-5G
苯并[b]噻吩-2-基硼酸 (contains varying amounts of Anhydride)
苯并[b]噻吩-2-基硼酸
分子式: C₈H₇BO₂S
溶点: 260°C
MDL编号: MFCD01075674
CAS号: 98437-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf5171597f186834ced4e6c0b36fe3fb3dca424&vid=6cf5171597f186834ced4e6c0b36fe3fb3dca424&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR