4-氟-2-甲基苯酚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 5-Fluoro-2-hydroxytoluene, 1-Fluoro-4-hydroxy-3-methylbenzene

F0387-25G F0387-5G
TCIAF0387-25GEA 0 CNY
TCIAF0387-25G TCIAF0387-5G
4-氟-2-甲基苯酚 ≥98.0% (by GC)
4-氟-2-甲基苯酚
分子式: C₇H₇FO
分子量: 126.13 g/mol
沸点: 93°C
溶点: 35
MDL编号: MFCD00075088
CAS号: 452-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c7c458b07cdac0bd0651cbea926b7c5c846b3e2&vid=a1b4e985e9d251637a6d1609301b38547a089c6c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 34.0 to 37.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR