4-乙烯基苯甲酸 ≥97.0% (by GC, titration analysis) stabilised

供应商: TCI
Warning

Synonyms: 对乙烯基苯甲酸

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

V0059-1G V0059-25G V0059-5G
TCIAV0059-1GEA 0 CNY
TCIAV0059-1G TCIAV0059-25G TCIAV0059-5G
4-乙烯基苯甲酸 ≥97.0% (by GC, titration analysis) stabilised
4-乙烯基苯甲酸
分子式: C₉H₈O₂
分子量: 148.16 g/mol
沸点: 282
溶点: 115
存储条件: Refrigerator
MDL编号: MFCD00002569
CAS号: 1075-49-6
EINECS: 214-053-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=191cf027d5252bf2c878f7f7061b794860a58743&vid=10701d78a96b76d66fafdea16d7e795e92178f3d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR