(2-甲基丙基)硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

I0597-25G I0597-5G
TCIAI0597-25GEA 0 CNY
TCIAI0597-25G TCIAI0597-5G
(2-甲基丙基)硼酸 (contains varying amounts of Anhydride)
(2-甲基丙基)硼酸
分子式: C₄H₁₁BO₂
MDL编号: MFCD00134156
CAS号: 84110-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bc6cc28ff1c940de5b8746fe7157aa490e8c433&vid=4bc6cc28ff1c940de5b8746fe7157aa490e8c433&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 121.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR