1-Propylboronic acid (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
Synonyms:
Propylboronic acid
P1942-1G
P1942-25G
P1942-5G
TCIAP1942-1GEA
0
CNY
TCIAP1942-1G
TCIAP1942-25G
TCIAP1942-5G
1-Propylboronic acid (contains varying amounts of Anhydride)
1-Propylboronic acid
分子式:
C₃H₉BO₂ 溶点: 106°C |
MDL编号:
MFCD01074564 CAS号: 17745-45-8 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 126.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...