1-Hexylboronic acid (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: Hexylboronic acid

H0913-1G H0913-5G
TCIAH0913-1GEA 0 CNY
TCIAH0913-1G TCIAH0913-5G
1-Hexylboronic acid (contains varying amounts of Anhydride)
1-Hexylboronic acid
分子式: C₆H₁₅BO₂
MDL编号: MFCD01074641
CAS号: 16343-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9365490736db6f59fe79c655d618c01d2ebda4fd&vid=9365490736db6f59fe79c655d618c01d2ebda4fd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 116.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR