Dipentyl ether ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Amyl ether, Di-n-amyl ether, n-Pentyl ether
A0451-25ML
A0451-500ML
TCIAA0451-25MLEA
0
CNY
TCIAA0451-25ML
TCIAA0451-500ML
Dipentyl ether ≥98.0% (by GC)
Dipentyl ether
Other Product Names / Synonyms: Diamyl Ether, Pentyl Ether, Dipentyl Ether
分子式:
C₁₀H₂₂O 沸点: 186°C 溶点: -69°C 闪点: 57°C |
MDL编号:
MFCD00009501 CAS号: 693-65-2 EINECS: 211-756-8 UN号: 3271 ADR: 3,III Merck Index: 14,00608 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...